Product Details

SNP ID
rs28624980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48028712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTCTGGGATTGTGCTATGGTGC[A/G]TGGAGGGTGAGGGAAGATGGGAAGA
Phenotype
MIM: 615526 MIM: 613788
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPZ2 PubMed Links

Gene Details

Gene
COPZ2
Gene Name
coatomer protein complex subunit zeta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016429.3 Intron NP_057513.1
XM_011524874.2 Intron XP_011523176.1
Gene
MIR152
Gene Name
microRNA 152
There are no transcripts associated with this gene.

View Full Product Details