Product Details

SNP ID
rs28384390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30116662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTACAGATTTTGAGGCAGCCGG[G/T]GGGCTCCGAAACAAAACAGAGGTCA
Phenotype
MIM: 616173
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EFCAB5 PubMed Links

Gene Details

Gene
EFCAB5
Gene Name
EF-hand calcium binding domain 5
There are no transcripts associated with this gene.

Gene
MIR3184
Gene Name
microRNA 3184
There are no transcripts associated with this gene.

Gene
MIR423
Gene Name
microRNA 423
There are no transcripts associated with this gene.

Gene
NSRP1
Gene Name
nuclear speckle splicing regulatory protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261467.1 217 Intron NP_001248396.1
NM_032141.3 217 Intron NP_115517.1
XM_011525345.2 217 Intron XP_011523647.1
XM_017025209.1 217 Intron XP_016880698.1
XM_017025210.1 217 UTR 5 XP_016880699.1
XM_017025211.1 217 Intron XP_016880700.1
XM_017025212.1 217 UTR 5 XP_016880701.1
XM_017025213.1 217 Intron XP_016880702.1
XM_017025214.1 217 Intron XP_016880703.1
XM_017025215.1 217 Intron XP_016880704.1

View Full Product Details