Product Details

SNP ID
rs28719851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:80109947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGAAACGCCCCGCCCGCTGGCC[C/G]GCCCGCCGCATCCCCGAGACCGGGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADNP2 PubMed Links

Gene Details

Gene
ADNP2
Gene Name
ADNP homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014913.3 222 Intron NP_055728.1
XM_005266656.4 222 UTR 5 XP_005266713.1
XM_011525883.2 222 Intron XP_011524185.1

View Full Product Details