Product Details
- SNP ID
-
rs28411237
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39519349 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCATCTGTAAGACCCTGGGGTAGG[A/G]CTGTGTTCAGTGAGTTTGAAAAATA
- Phenotype
-
MIM: 607919
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SELV
PubMed Links
Gene Details
- Gene
- SELV
- Gene Name
- selenoprotein V
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182704.1 |
|
Intron |
|
|
NP_874363.1 |
View Full Product Details