Product Details

SNP ID
rs28699037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42276077 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGGCTTCTTGGAAGAAGGGCCA[G/T]ATGCCTATGAACTGAAACCTGAAGG
Phenotype
MIM: 612082
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CIC PubMed Links

Gene Details

Gene
CIC
Gene Name
capicua transcriptional repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304815.1 Intron NP_001291744.1
NM_015125.4 Intron NP_055940.3
XM_005258673.2 Intron XP_005258730.1
XM_005258674.2 Intron XP_005258731.1
XM_005258675.2 Intron XP_005258732.1
XM_011526660.2 Intron XP_011524962.1
XM_011526661.2 Intron XP_011524963.1
XM_011526662.2 Intron XP_011524964.1
XM_011526663.2 Intron XP_011524965.1
XM_011526664.2 Intron XP_011524966.1
XM_011526665.2 Intron XP_011524967.1
XM_011526666.2 Intron XP_011524968.1
XM_011526667.2 Intron XP_011524969.1

View Full Product Details