Product Details

SNP ID
rs8100301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42728571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAAATTCAGGACAGGCCACCAGG[A/G]CTCTTTCTCCACAGATGCTGGTCCC
Phenotype
MIM: 176392
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSG3 PubMed Links

Gene Details

Gene
PSG3
Gene Name
pregnancy specific beta-1-glycoprotein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021016.3 Intron NP_066296.2
XM_011527126.1 Intron XP_011525428.1
XM_011527127.1 Intron XP_011525429.1

View Full Product Details