Product Details

SNP ID
rs35071847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45039617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTTGCAACACTCCCTCCCCGTG[G/T]TCCCACTCCCTCTAGCATCTTCATC
Phenotype
MIM: 604758
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLASRP PubMed Links

Gene Details

Gene
CLASRP
Gene Name
CLK4 associating serine/arginine rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278439.1 Intron NP_001265368.1
NM_007056.2 Intron NP_008987.2
XM_011526396.2 Intron XP_011524698.1
XM_011526397.2 Intron XP_011524699.1
Gene
RELB
Gene Name
RELB proto-oncogene, NF-kB subunit
There are no transcripts associated with this gene.

View Full Product Details