Product Details

SNP ID
rs34383186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45382459 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCCTGTTGCCTCTTCTTTTTCC[C/T]GTGGGATCCCCCTTTTCCCCAACCC
Phenotype
MIM: 607463
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1R13L PubMed Links

Gene Details

Gene
PPP1R13L
Gene Name
protein phosphatase 1 regulatory subunit 13 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142502.1 Intron NP_001135974.1
NM_006663.3 Intron NP_006654.2
XM_017026177.1 Intron XP_016881666.1
XM_017026178.1 Intron XP_016881667.1
XM_017026179.1 Intron XP_016881668.1

View Full Product Details