Product Details

SNP ID
rs28379613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51452378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAGAGGGTTCTTGTTCTATGAGA[A/T]TCAGCCTCTGACTTCTTTGCTGGAG
Phenotype
MIM: 605639
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SIGLEC8 PubMed Links

Gene Details

Gene
SIGLEC8
Gene Name
sialic acid binding Ig like lectin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014442.2 1529 UTR 3 NP_055257.2
XM_011526734.2 1529 UTR 3 XP_011525036.1
XM_011526735.2 1529 UTR 3 XP_011525037.1

View Full Product Details