Product Details

SNP ID
rs1130472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54830101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCAGCATTTCCCTCTCTCCAGGA[C/T]TCTGATGAACAAGACCCTGAGGAGG
Phenotype
MIM: 604955 MIM: 604946
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIR2DS4 PubMed Links

Gene Details

Gene
KIR2DS4
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4
There are no transcripts associated with this gene.

Gene
KIR3DL1
Gene Name
killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322168.1 1224 Silent Mutation GAC,GAT D387D NP_001309097.1
NM_013289.2 1224 Silent Mutation GAC,GAT D387D NP_037421.2

View Full Product Details