Product Details

SNP ID
rs28498001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:31582026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAAAAAAAAAAAGTTAGCGGC[A/C]TGTGGTGGCTTACGTTGTATTCCCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927277 PubMed Links

Gene Details

Gene
LOC101927277
Gene Name
uncharacterized LOC101927277
There are no transcripts associated with this gene.

Gene
THEG5
Gene Name
testis highly expressed protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205273.2 Intron NP_001192202.1
NM_001278575.1 Intron NP_001265504.1
NM_001278576.1 Intron NP_001265505.1
NM_001278577.1 Intron NP_001265506.1
NM_001278578.1 Intron NP_001265507.1
XM_011526302.2 Intron XP_011524604.1
XM_011526303.2 Intron XP_011524605.1
XM_017026126.1 Intron XP_016881615.1
XM_017026127.1 Intron XP_016881616.1
XM_017026128.1 Intron XP_016881617.1
XM_017026129.1 Intron XP_016881618.1
XM_017026130.1 Intron XP_016881619.1
XM_017026131.1 Intron XP_016881620.1
XM_017026132.1 Intron XP_016881621.1
XM_017026133.1 Intron XP_016881622.1

View Full Product Details