Product Details
- SNP ID
-
rs28758984
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:1065965 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTCCCGGAGCACAGGCCAGGGCC[G/T]GGCCAGAAGCCCATCCGACATCCAG
- Phenotype
-
MIM: 605414
MIM: 601155
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ABCA7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148921927] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABCA7
- Gene Name
- ATP binding cassette subfamily A member 7
- Gene
- ARHGAP45
- Gene Name
- Rho GTPase activating protein 45
View Full Product Details