Product Details

SNP ID
rs28670427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21806816 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTAGTGTAATGTCTGAAGTGTC[C/T]GCGCCTTAGATATTTCTACTGTGAA
Phenotype
MIM: 603972
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF43 PubMed Links

Gene Details

Gene
ZNF43
Gene Name
zinc finger protein 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256648.1 9049 Intron NP_001243577.1
NM_001256649.1 9049 UTR 3 NP_001243578.1
NM_001256650.1 9049 Intron NP_001243579.1
NM_001256651.1 9049 Intron NP_001243580.1
NM_001256653.1 9049 Intron NP_001243582.1
NM_001256654.1 9049 UTR 3 NP_001243583.1
NM_003423.3 9049 UTR 3 NP_003414.2
XM_011528257.2 9049 Intron XP_011526559.1
XM_011528259.2 9049 UTR 3 XP_011526561.1
XM_017027207.1 9049 Intron XP_016882696.1
XM_017027208.1 9049 Intron XP_016882697.1
XM_017027209.1 9049 Intron XP_016882698.1
XM_017027210.1 9049 Intron XP_016882699.1
XM_017027211.1 9049 UTR 3 XP_016882700.1
XM_017027212.1 9049 Intron XP_016882701.1
XM_017027213.1 9049 Intron XP_016882702.1
XM_017027214.1 9049 Intron XP_016882703.1
XM_017027215.1 9049 Intron XP_016882704.1
XM_017027216.1 9049 Intron XP_016882705.1

View Full Product Details