Product Details

SNP ID
rs28565143
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10106787 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTAAGTCTCCCCAGCTGGAAAAA[A/G]AGACCCTCATGGGGAGCATCATTTC
Phenotype
MIM: 609336 MIM: 603913 MIM: 602697 MIM: 607793
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ANGPTL6 PubMed Links
Additional Information
For this assay, SNP(s) [rs3841592] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ANGPTL6
Gene Name
angiopoietin like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321411.1 Intron NP_001308340.1
NM_031917.2 Intron NP_114123.2
XM_005260091.3 Intron XP_005260148.1
XM_011528347.2 Intron XP_011526649.1
XM_011528348.2 Intron XP_011526650.1
XM_011528349.2 Intron XP_011526651.1
XM_011528350.2 Intron XP_011526652.1
XM_017027347.1 Intron XP_016882836.1
Gene
EIF3G
Gene Name
eukaryotic translation initiation factor 3 subunit G
There are no transcripts associated with this gene.

Gene
P2RY11
Gene Name
purinergic receptor P2Y11
There are no transcripts associated with this gene.

Gene
PPAN
Gene Name
peter pan homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020230.5 Intron NP_064615.3
Gene
PPAN-P2RY11
Gene Name
PPAN-P2RY11 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040664.2 Intron NP_001035754.1
NM_001198690.1 Intron NP_001185619.1
Gene
SNORD105
Gene Name
small nucleolar RNA, C/D box 105
There are no transcripts associated with this gene.

Gene
SNORD105B
Gene Name
small nucleolar RNA, C/D box 105B
There are no transcripts associated with this gene.

View Full Product Details