Product Details

SNP ID
rs35654716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10289752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATGACGCGAGAGAGCGTGGTCG[C/T]CCCCAGAAGGCCCTGGGCCATCATG
Phenotype
MIM: 147840 MIM: 614088 MIM: 601852
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ICAM1 PubMed Links

Gene Details

Gene
ICAM1
Gene Name
intercellular adhesion molecule 1
There are no transcripts associated with this gene.

Gene
ICAM4
Gene Name
intercellular adhesion molecule 4 (Landsteiner-Wiener blood group)
There are no transcripts associated with this gene.

Gene
ICAM5
Gene Name
intercellular adhesion molecule 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003259.3 Intron NP_003250.3
XM_011528229.1 Intron XP_011526531.1
XM_017027185.1 Intron XP_016882674.1

View Full Product Details