Product Details

SNP ID
rs35889404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:41140520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACTGTGCTACCTTCAGGGTGTT[A/G]GTAGACCTGAGGGAGAGACAGTGTG
Phenotype
MIM: 172420
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLCG1 PubMed Links

Gene Details

Gene
PLCG1
Gene Name
phospholipase C gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002660.2 Intron NP_002651.2
NM_182811.1 Intron NP_877963.1
XM_005260438.2 Intron XP_005260495.1
XM_011528865.1 Intron XP_011527167.1
XM_011528867.2 Intron XP_011527169.1
Gene
PLCG1-AS1
Gene Name
PLCG1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details