Product Details

SNP ID
rs36105541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:6770355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGCAGGGACGCCGTGGTGCCC[C/T]CCTACATGCTAGACCTGTATCGCAG
Phenotype
MIM: 112261
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BMP2 PubMed Links

Gene Details

Gene
BMP2
Gene Name
bone morphogenetic protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001200.3 1014 Missense Mutation CCC,TCC P77S NP_001191.1
XM_011529323.2 1014 Intron XP_011527625.1

View Full Product Details