Product Details

SNP ID
rs17883270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:31661769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCCTGAGCAGCTTGCATAGTTC[C/G]TTCTTTTAAGCGCCTGACTTCGTTT
Phenotype
MIM: 616023 MIM: 147450
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC102724449 PubMed Links

Gene Details

Gene
LOC102724449
Gene Name
uncharacterized LOC102724449
There are no transcripts associated with this gene.

Gene
SCAF4
Gene Name
SR-related CTD associated factor 4
There are no transcripts associated with this gene.

Gene
SOD1
Gene Name
superoxide dismutase 1, soluble
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000454.4 Intron NP_000445.1

View Full Product Details