Product Details

SNP ID
rs17880191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33407303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCACCGAGAAAGACAGTAAGTCC[C/T]CAGCAACTTAAAAACCATTGCATGT
Phenotype
MIM: 147569
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFNGR2 PubMed Links

Gene Details

Gene
IFNGR2
Gene Name
interferon gamma receptor 2 (interferon gamma transducer 1)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005534.3 Intron NP_005525.2
XM_005260969.2 Intron XP_005261026.1
XM_011529553.1 Intron XP_011527855.1
XM_011529554.2 Intron XP_011527856.1

View Full Product Details