Product Details

SNP ID
rs28593087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19445149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTAGTACAGAATATTAAAGAT[C/T]CCCAAATGTCCATTCCCAATGCGGC
Phenotype
MIM: 605089 MIM: 601754
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C22orf39 PubMed Links

Gene Details

Gene
C22orf39
Gene Name
chromosome 22 open reading frame 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166242.1 Intron NP_001159714.1
NM_173793.4 Intron NP_776154.3
Gene
MRPL40
Gene Name
mitochondrial ribosomal protein L40
There are no transcripts associated with this gene.

Gene
UFD1L
Gene Name
ubiquitin fusion degradation 1 like (yeast)
There are no transcripts associated with this gene.

View Full Product Details