Product Details

SNP ID
rs28626693
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37023889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGAAGCCAGCAGGGCCTTGCCC[G/T]AGGATACCTCCACCCACCCTCCTGC
Phenotype
MIM: 602496 MIM: 180370
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MPST PubMed Links

Gene Details

Gene
MPST
Gene Name
mercaptopyruvate sulfurtransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013436.2 271 Intron NP_001013454.1
NM_001130517.2 271 UTR 5 NP_001123989.1
NM_021126.5 271 Intron NP_066949.2
XM_005261610.3 271 UTR 5 XP_005261667.1
XM_011530195.1 271 Intron XP_011528497.1
XM_011530196.1 271 Intron XP_011528498.1
Gene
TST
Gene Name
thiosulfate sulfurtransferase
There are no transcripts associated with this gene.

View Full Product Details