Product Details

SNP ID
rs28566067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40045899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGACTTTAAGTTCGTGCTTTCTC[A/C]TGTGTTTTGGCTGTTTTATTACTTT
Phenotype
MIM: 610740
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC100130899 PubMed Links

Gene Details

Gene
LOC100130899
Gene Name
uncharacterized LOC100130899
There are no transcripts associated with this gene.

Gene
TNRC6B
Gene Name
trinucleotide repeat containing 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024843.1 Intron NP_001020014.1
NM_001162501.1 Intron NP_001155973.1
NM_015088.2 Intron NP_055903.2

View Full Product Details