Product Details

SNP ID
rs28407826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41841046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTCTGTAAAGGTGTGTCTAATG[A/G]GGGAAGGAAAGCACCTCAAGTTCAG
Phenotype
MIM: 600481
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC134 PubMed Links

Gene Details

Gene
CCDC134
Gene Name
coiled-coil domain containing 134
There are no transcripts associated with this gene.

Gene
SREBF2
Gene Name
sterol regulatory element binding transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004599.3 Intron NP_004590.2
XM_006724310.2 Intron XP_006724373.1
XM_011530347.1 Intron XP_011528649.1
XM_017028921.1 Intron XP_016884410.1
XM_017028922.1 Intron XP_016884411.1

View Full Product Details