Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001318038.1 | 1847 | UTR 3 | NP_001304967.1 | ||
NM_017436.5 | 1847 | UTR 3 | NP_059132.1 | ||
XM_005261644.2 | 1847 | UTR 3 | XP_005261701.1 | ||
XM_005261646.4 | 1847 | UTR 3 | XP_005261703.1 | ||
XM_005261647.2 | 1847 | UTR 3 | XP_005261704.1 | ||
XM_005261648.3 | 1847 | UTR 3 | XP_005261705.1 | ||
XM_006724265.3 | 1847 | UTR 3 | XP_006724328.1 | ||
XM_006724266.3 | 1847 | UTR 3 | XP_006724329.1 | ||
XM_011530233.2 | 1847 | UTR 3 | XP_011528535.1 | ||
XM_017028831.1 | 1847 | UTR 3 | XP_016884320.1 |