Product Details

SNP ID
rs28694368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23361212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCCCTCCACCAAGGGGGGCCTG[A/G]GGGATCTCTGCCTGTAGTGTTTCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CES5AP1 PubMed Links

Gene Details

Gene
CES5AP1
Gene Name
carboxylesterase 5A pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details