Product Details

SNP ID
rs34174598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23790088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACCTGGGTCCCAAGAGGGTGAAG[C/G]CTGGTTCAGAGGAAAGCCAGGAAGG
Phenotype
MIM: 185261 MIM: 601607
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MMP11 PubMed Links

Gene Details

Gene
MMP11
Gene Name
matrix metallopeptidase 11
There are no transcripts associated with this gene.

Gene
SMARCB1
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007468.2 Intron NP_001007469.1
NM_001317946.1 Intron NP_001304875.1
NM_003073.4 Intron NP_003064.2
XM_011530345.2 Intron XP_011528647.1

View Full Product Details