Product Details

SNP ID
rs35961970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23836352 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGCTAGATTGGCATCAGCCTGA[A/T]GGCACCACTGGCAGGAACATCTGTA
Phenotype
MIM: 610305 MIM: 601607
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DERL3 PubMed Links

Gene Details

Gene
DERL3
Gene Name
derlin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002862.2 Intron NP_001002862.1
NM_001135751.1 Intron NP_001129223.1
NM_198440.3 Intron NP_940842.2
XM_011530505.2 Intron XP_011528807.1
XM_011530506.2 Intron XP_011528808.1
XM_017029078.1 Intron XP_016884567.1
XM_017029079.1 Intron XP_016884568.1
XM_017029080.1 Intron XP_016884569.1
XM_017029081.1 Intron XP_016884570.1
XM_017029082.1 Intron XP_016884571.1
Gene
SMARCB1
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1
There are no transcripts associated with this gene.

View Full Product Details