Product Details

SNP ID
rs35468138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23894665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGCTGGGGAGGCGACTCCTGAA[C/T]GGAGCTGGGGGGCGGGGCGGGGGGA
Phenotype
MIM: 153620 MIM: 610367
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIF PubMed Links

Gene Details

Gene
MIF
Gene Name
macrophage migration inhibitory factor (glycosylation-inhibiting factor)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002415.1 Intron NP_002406.1
Gene
MIF-AS1
Gene Name
MIF antisense RNA 1
There are no transcripts associated with this gene.

Gene
SLC2A11
Gene Name
solute carrier family 2 member 11
There are no transcripts associated with this gene.

View Full Product Details