Product Details

SNP ID
rs28470453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24222518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGGATCAAAGACTCTCCTCGCC[C/T]TAGTCCCTGGGCCTTAGCTGTGAAG
Phenotype
MIM: 137168
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GGT5 PubMed Links

Gene Details

Gene
GGT5
Gene Name
gamma-glutamyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099781.2 Intron NP_001093251.1
NM_001099782.2 Intron NP_001093252.1
NM_001302464.1 Intron NP_001289393.1
NM_001302465.1 Intron NP_001289394.1
NM_004121.3 Intron NP_004112.2
XM_005261557.2 Intron XP_005261614.1
XM_005261558.2 Intron XP_005261615.1
XM_011530133.1 Intron XP_011528435.1
XM_011530134.1 Intron XP_011528436.1
XM_011530135.1 Intron XP_011528437.1
XM_011530136.1 Intron XP_011528438.1
XM_011530137.2 Intron XP_011528439.1
XM_017028768.1 Intron XP_016884257.1
XM_017028769.1 Intron XP_016884258.1

View Full Product Details