Product Details
- SNP ID
-
rs28718659
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:46340043 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTGGAGATTTGGACTTCATGGGG[A/G]GGGAGTGTGGGAGTGGAAGGTGAGG
- Phenotype
-
MIM: 607477
MIM: 610230
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GTSE1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112511097] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GTSE1
- Gene Name
- G2 and S-phase expressed 1
There are no transcripts associated with this gene.
- Gene
- TRMU
- Gene Name
- tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase
View Full Product Details