Product Details

SNP ID
rs35149516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:54445940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAATATATTTTTTAAAAGCCCAC[A/G]TTTTGTGAAAAGATGAATTAAAAAA
Phenotype
MIM: 300546 MIM: 300945
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGD1 PubMed Links

Gene Details

Gene
FGD1
Gene Name
FYVE, RhoGEF and PH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004463.2 3789 UTR 3 NP_004454.2
Gene
TSR2
Gene Name
TSR2, ribosome maturation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058163.1 3789 Intron NP_477511.1

View Full Product Details