Product Details

SNP ID
rs28382681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:55002528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATGTAAACATACATGCACCGTG[A/G]AGAAGTTCCCAAATTTGCCTTTCCT
Phenotype
MIM: 301300 MIM: 300773 MIM: 311790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALAS2 PubMed Links

Gene Details

Gene
ALAS2
Gene Name
5'-aminolevulinate synthase 2
There are no transcripts associated with this gene.

Gene
APEX2
Gene Name
apurinic/apyrimidinic endodeoxyribonuclease 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271748.1 Intron NP_001258677.1
NM_014481.3 Intron NP_055296.2
Gene
PFKFB1
Gene Name
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1
There are no transcripts associated with this gene.

View Full Product Details