Product Details

SNP ID
rs28589992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:57122921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTCATATCTAGTGCTGATGCAA[C/G]TGAAAGTTTATACAATTTTTATGGA
Phenotype
MIM: 300654 MIM: 300517
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAAH2 PubMed Links

Gene Details

Gene
FAAH2
Gene Name
fatty acid amide hydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174912.3 Intron NP_777572.2
XM_005261982.1 Intron XP_005262039.1
XM_005261985.2 Intron XP_005262042.1
XM_005261986.3 Intron XP_005262043.1
XM_005261987.3 Intron XP_005262044.1
XM_011530767.2 Intron XP_011529069.1
XM_017029290.1 Intron XP_016884779.1
XM_017029291.1 Intron XP_016884780.1
XM_017029292.1 Intron XP_016884781.1
XM_017029293.1 Intron XP_016884782.1
XM_017029294.1 Intron XP_016884783.1
Gene
SPIN2B
Gene Name
spindlin family member 2B
There are no transcripts associated with this gene.

View Full Product Details