Product Details
- SNP ID
-
rs28442045
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:153765792 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGGCCTTCCTCCAGGCGCTGCTC[C/T]GCCCTGAGTCGCAGCGGCCCCACTC
- Phenotype
-
MIM: 300214
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLXNB3
PubMed Links
Gene Details
- Gene
- PLXNB3
- Gene Name
- plexin B3
View Full Product Details