Product Details

SNP ID
rs11538409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154478737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCGGAAAGGATACAATATGTGC[A/G]GCCGCATTCGTGACCCCCTGGCCGC
Phenotype
MIM: 300060 MIM: 300022 MIM: 312090 MIM: 312070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAGE3 PubMed Links

Gene Details

Gene
LAGE3
Gene Name
L antigen family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006014.4 521 Missense Mutation CCG,CTG P60L NP_006005.2
Gene
PLXNA3
Gene Name
plexin A3
There are no transcripts associated with this gene.

Gene
SLC10A3
Gene Name
solute carrier family 10 member 3
There are no transcripts associated with this gene.

Gene
UBL4A
Gene Name
ubiquitin like 4A
There are no transcripts associated with this gene.

View Full Product Details