Product Details
- SNP ID
-
hCV61427343
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:13783501 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAAAGGACGCAATTCCATAGATG[A/T]CATACTGCATCAGTTGTATCCTACA
- Phenotype
-
MIM: 300051
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GPM6B
PubMed Links
Gene Details
- Gene
- GPM6B
- Gene Name
- glycoprotein M6B
There are no transcripts associated with this gene.
View Full Product Details