Product Details

SNP ID
rs28655098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:132216951 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAACCCAGAAGTCGATGTCGGA[G/T]TACCACACCCCCTTCTAACAAATTA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RAP2C PubMed Links

Gene Details

Gene
RAP2C
Gene Name
RAP2C, member of RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271186.1 Intron NP_001258115.1
NM_001271187.1 Intron NP_001258116.1
NM_021183.4 Intron NP_067006.3
XM_006724775.2 Intron XP_006724838.1
XM_011531376.1 Intron XP_011529678.1
Gene
RAP2C-AS1
Gene Name
RAP2C antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details