Product Details

SNP ID
rs28490493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:141616896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTCATTGACTTTTCAATGAACA[A/G]TTTTCTTATAGGTAGCCAAAGAGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC645188 PubMed Links

Gene Details

Gene
LOC645188
Gene Name
uncharacterized LOC645188
There are no transcripts associated with this gene.

Gene
SPANXA2-OT1
Gene Name
SPANXA2 overlapping transcript 1
There are no transcripts associated with this gene.

View Full Product Details