Product Details

SNP ID
rs35567891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:9610745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCATGAAAACAGGAATAAAAATC[A/G]CCACAGTGTCTTGAGTAACTCTCCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBMY3AP PubMed Links

Gene Details

Gene
RBMY3AP
Gene Name
RNA binding motif protein, Y-linked, family 3, member A pseudogene
There are no transcripts associated with this gene.

View Full Product Details