Product Details

SNP ID
rs34820858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:6868771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCTGGTACCACTTCAAAGGGGT[G/T]AGCACCTTAAGAGAAAGATATATTA
Phenotype
MIM: 410000
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AMELY PubMed Links

Gene Details

Gene
AMELY
Gene Name
amelogenin, Y-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143.1 247 Intron NP_001134.1
XM_017030042.1 247 Silent Mutation CTA,CTC L50L XP_016885531.1

View Full Product Details