Product Details

SNP ID
rs28380626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:88382653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAATTGAATGTTTTTAAGAGTCA[A/G]TTTGAACATTCAGCAGTTCATGAAT
Phenotype
MIM: 609249
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HERC6 PubMed Links

Gene Details

Gene
HERC6
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165136.1 Intron NP_001158608.1
NM_017912.3 Intron NP_060382.3
XM_005263083.3 Intron XP_005263140.1
XM_011532053.2 Intron XP_011530355.1
XM_017008333.1 Intron XP_016863822.1

View Full Product Details