Product Details

SNP ID
rs35270074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:89726976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGTCTGGTTCTCAGTTTCTTCA[C/T]CTGTAAAATGTGAGAACAAAATCAA
Phenotype
MIM: 163890
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNCA PubMed Links

Gene Details

Gene
SNCA
Gene Name
synuclein alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000345.3 Intron NP_000336.1
NM_001146054.1 Intron NP_001139526.1
NM_001146055.1 Intron NP_001139527.1
NM_007308.2 Intron NP_009292.1
XM_011532203.1 Intron XP_011530505.1
XM_011532204.2 Intron XP_011530506.1
XM_011532205.2 Intron XP_011530507.1
XM_011532206.1 Intron XP_011530508.1
XM_011532207.1 Intron XP_011530509.1
XM_011532208.2 Intron XP_011530510.1
XM_017008562.1 Intron XP_016864051.1
XM_017008563.1 Intron XP_016864052.1

View Full Product Details