Product Details

SNP ID
rs28485130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76953879 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAATTGATTAAAAGAACAAAACTG[C/T]CCATAACCCACTATCCTGATATAAT
Phenotype
MIM: 612887
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEPT11 PubMed Links

Gene Details

Gene
SEPT11
Gene Name
septin 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306147.1 Intron NP_001293076.1
NM_018243.3 Intron NP_060713.1
XM_006714259.3 Intron XP_006714322.1
XM_017008400.1 Intron XP_016863889.1
XM_017008401.1 Intron XP_016863890.1
XM_017008402.1 Intron XP_016863891.1
XM_017008403.1 Intron XP_016863892.1
XM_017008404.1 Intron XP_016863893.1

View Full Product Details