Product Details

SNP ID
rs28730635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99074196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTGATTTTTTTTCTTTAGGAGG[A/G]GGATGAAATATAAATATATATTTTT
Phenotype
MIM: 103710
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADH5 PubMed Links

Gene Details

Gene
ADH5
Gene Name
alcohol dehydrogenase 5 (class III), chi polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000671.4 Intron NP_000662.3

View Full Product Details