Product Details

SNP ID
rs29001230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99125547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCACTTCAACTTCTATCTCCTAA[A/C]TTGAGGAGTATTCCTTATGGGGATA
Phenotype
MIM: 103740
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADH4 PubMed Links

Gene Details

Gene
ADH4
Gene Name
alcohol dehydrogenase 4 (class II), pi polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000670.4 Intron NP_000661.2
NM_001306171.1 Intron NP_001293100.1
NM_001306172.1 Intron NP_001293101.1
XM_017007713.1 Intron XP_016863202.1
XM_017007714.1 Intron XP_016863203.1
XM_017007715.1 Intron XP_016863204.1
Gene
LOC100507053
Gene Name
uncharacterized LOC100507053
There are no transcripts associated with this gene.

View Full Product Details