Product Details

SNP ID
rs28914787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99308117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCCTGTCATTTCAATTCTACATT[A/G]ATTCTACATTAGCATCTTCAAAAAA
Phenotype
MIM: 103720
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADH1B PubMed Links

Gene Details

Gene
ADH1B
Gene Name
alcohol dehydrogenase 1B (class I), beta polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000668.5 Intron NP_000659.2
NM_001286650.1 Intron NP_001273579.1
Gene
LOC100507053
Gene Name
uncharacterized LOC100507053
There are no transcripts associated with this gene.

View Full Product Details