Product Details

SNP ID
rs28734872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:105902550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTGAGGTGAGAATGCAGAATCT[A/G]TTTTTTGTTTGTTTGCTTTTAGGTA
Phenotype
MIM: 610306
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NPNT PubMed Links

Gene Details

Gene
NPNT
Gene Name
nephronectin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033047.2 Intron NP_001028219.1
NM_001184690.1 Intron NP_001171619.1
NM_001184691.1 Intron NP_001171620.1
NM_001184692.1 Intron NP_001171621.1
NM_001184693.1 Intron NP_001171622.1
XM_005262888.4 Intron XP_005262945.1
XM_005262890.1 Intron XP_005262947.1
XM_011531820.2 Intron XP_011530122.1
XM_011531822.2 Intron XP_011530124.1
XM_011531823.2 Intron XP_011530125.1
XM_011531824.2 Intron XP_011530126.1
XM_011531825.2 Intron XP_011530127.1
XM_017007984.1 Intron XP_016863473.1

View Full Product Details