Product Details

SNP ID
rs28854838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:118693090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTTGTGTTTCCAAGCAGCTGCA[C/T]GATTTTACATTTTTACTAACAACGT
Phenotype
MIM: 616504
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929741 PubMed Links

Gene Details

Gene
LOC101929741
Gene Name
uncharacterized LOC101929741
There are no transcripts associated with this gene.

Gene
METTL14
Gene Name
methyltransferase like 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020961.3 Intron NP_066012.1
XM_005263166.4 Intron XP_005263223.1
XM_011532162.2 Intron XP_011530464.1

View Full Product Details