Product Details
- SNP ID
-
rs28405657
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:119138735 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTGCATCAGAATTTTTTTTCAGC[A/C]TCATAACTCATTTAATTTTTATTAC
- Phenotype
-
MIM: 605602
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MYOZ2
PubMed Links
Gene Details
- Gene
- MYOZ2
- Gene Name
- myozenin 2
View Full Product Details