Product Details

SNP ID
rs28624232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:128819244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGGCTGTGGGTTGTTTTTGTTT[C/T]GCTCTTTTTGCCCAGGCTGGAGTGC
Phenotype
MIM: 610514
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JADE1 PubMed Links

Gene Details

Gene
JADE1
Gene Name
jade family PHD finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287437.1 Intron NP_001274366.1
NM_001287439.1 Intron NP_001274368.1
NM_001287440.1 Intron NP_001274369.1
NM_001287441.1 Intron NP_001274370.1
NM_001287442.1 Intron NP_001274371.1
NM_001287443.1 Intron NP_001274372.1
NM_024900.4 Intron NP_079176.2
NM_199320.3 Intron NP_955352.1
XM_005263232.1 Intron XP_005263289.1
XM_017008626.1 Intron XP_016864115.1
XM_017008627.1 Intron XP_016864116.1

View Full Product Details